Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fry
Entrez Gene ID
39122
Gene Name
furry
Gene Aliases
CG14171, CG32045, CG6774, CG6780, Dmel\CG32045, Dmel_CG32045, Fry, RE-B, SS3-7, l(3)02240, yeti
Chromosome Location
Chr.3L: 9631834 - 9679103 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016081
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259770.1 NP_001246699.1 16 - 17 5906 84
NM_001259771.1 NP_001246700.1 15 - 16 5403 84
NM_001259772.1 NP_001246701.1 16 - 17 5547 84
NM_001274708.1 NP_001261637.1 15 - 16 5762 84
NM_001274709.1 NP_001261638.1 16 - 17 5547 84
NM_001274710.1 NP_001261639.1 15 - 16 5403 84
NM_168356.2 NP_729519.2 16 - 17 5547 84
NM_168357.3 NP_729520.1 15 - 16 5403 84

View Full Product Details