Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Shark
- Entrez Gene ID
-
44353
- Gene Name
- SH2 ankyrin repeat kinase
- Gene Aliases
-
CG18247, Dmel\CG18247, Dmel_CG18247, Dtk7, SHARK, SYK/SHARK, Syk, Tk7, dtk7, l(2)W4, l(2R)W4, shark
- Chromosome Location
-
Chr.2R: 16199426 - 16202884 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015295
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080004.4 |
NP_524743.2 |
4 - 5 |
2383 |
76 |
GenBank mRNA |
AY051937.1 |
- |
4 - 5 |
2358 |
76 |
View Full Product Details