Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
X11L
Entrez Gene ID
252671
Gene Name
CG5675-PA
Gene Aliases
CG5675, CG5678, Dmel\CG5675, Dmel_CG5675, Dmint, Dmint1, MINT, Mint, X11, X11-like, X11/Mint, X11Lalpha, dX11, dX11/Mint, dX11L, dX11Lalpha, x11/Mint, x11Lalpha
Chromosome Location
Chr.X: 17599608 - 17606879 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026313
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169310.2 NP_001162781.1 5 - 6 2198 62
NM_001272725.2 NP_001259654.1 5 - 6 2557 62
NM_001272726.2 NP_001259655.2 5 - 6 2198 62
NM_001298447.1 NP_001285376.1 5 - 6 2198 62
NM_132996.4 NP_573224.2 5 - 6 2195 62
GenBank mRNA AY051760.1 - 5 - 6 2162 62

View Full Product Details