Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gdap1
Entrez Gene ID
38597
Gene Name
CG4623-PA
Gene Aliases
CG4623, Dmel\CG4623, Dmel_CG4623, GDAP1
Chromosome Location
Chr.3L: 5141953 - 5144317 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035587
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300010.1 NP_001286939.1 2 - 3 668 70
NM_139688.4 NP_647945.1 2 - 3 668 70
GenBank mRNA AY071643.1 - 2 - 3 666 70

View Full Product Details