Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir64a
- Entrez Gene ID
-
38616
- Gene Name
- Ionotropic receptor 64a
- Gene Aliases
-
CG10633, CT29782, DmIr64a, DmelIR64a, Dmel\CG10633, Dmel_CG10633, IR64a, Ir64, ir64a
- Chromosome Location
-
Chr.3L: 5378786 - 5382821 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035604
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139705.1 |
NP_647962.1 |
4 - 5 |
1949 |
103 |
View Full Product Details