Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hsf
Entrez Gene ID
37068
Gene Name
Heat shock factor
Gene Aliases
CG5748, D-HSF, Dm-Hsf, DmHSF, DmHSFa, Dmel\CG5748, Dmel_CG5748, HSF, HSF1, HSFa, Hsf1, dHSF, dHSFa, heat shock factor, hsf, l(2)03091
Chromosome Location
Chr.2R: 18130303 - 18134634 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001222
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043092.3 NP_001036557.1 5 - 6 2165 76
NM_057227.5 NP_476575.1 6 - 7 2093 76
GenBank mRNA AB211168.1 - 4 - 5 1922 76
M60070.1 - 6 - 7 2078 76

View Full Product Details