Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- vnd
- Entrez Gene ID
-
31003
- Gene Name
- ventral nervous system defective
- Gene Aliases
-
CG6172, Dm vnd, Dm-NK2, Dmel\CG6172, Dmel_CG6172, EC6, EG:118B3.1, HDC16822, NK-2, NK2, Nkx2, VND, Vnd, dNK-2, l(1)1Bf, l(1)EA142, l(1)EC6, l(1)GA100, l(1)GA122, l(1)RC24, l(1)VA208, l(1)VE769, lab, nk-2, vnd/NK-2
- Chromosome Location
-
Chr.X: 582121 - 598151 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261930
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057438.4 |
NP_476786.2 |
1 - 2 |
913 |
51 |
View Full Product Details