Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ine
Entrez Gene ID
33659
Gene Name
inebriated
Gene Aliases
CG15444, CG15444d, DMINE, Dmel\CG15444, Dmel_CG15444, ine/rosA, rosA
Chromosome Location
Chr.2L: 4468000 - 4477022 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011603
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169400.2 NP_001162871.1 7 - 8 1969 66
NM_057664.6 NP_477012.1 7 - 8 2167 66
NM_057665.5 NP_477013.1 5 - 6 775 66
NM_175958.3 NP_787972.1 7 - 8 1955 66
NM_175959.3 NP_787973.1 7 - 8 1991 66
GenBank mRNA AY128432.1 - 7 - 8 1971 66
BT009955.1 - 7 - 8 1993 66
BT099793.1 - 5 - 6 758 66
U66460.1 - 5 - 6 734 66

View Full Product Details