Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ine
Entrez Gene ID
33659
Gene Name
inebriated
Gene Aliases
CG15444, CG15444d, DMINE, Dmel\CG15444, Dmel_CG15444, ine/rosA, rosA
Chromosome Location
Chr.2L: 4468000 - 4477022 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011603
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169400.2 NP_001162871.1 8 - 9 2925 79
NM_057664.6 NP_477012.1 8 - 9 3123 79
NM_057665.5 NP_477013.1 6 - 7 1731 79
NM_175958.3 NP_787972.1 8 - 9 2911 79
NM_175959.3 NP_787973.1 8 - 9 2947 79
GenBank mRNA AY128432.1 - 8 - 9 2927 79
BT009955.1 - 8 - 9 2949 79
BT099793.1 - 6 - 7 1714 79

View Full Product Details