Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bro
Entrez Gene ID
38202
Gene Name
Brother
Gene Aliases
CG7960, Dmel\CG7960, Dmel_CG7960, bro, rpbeta1
Chromosome Location
Chr.3L: 1647869 - 1648612 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013755
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057718.3 NP_477066.2 1 - 1 645 63

View Full Product Details