Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hr38
Entrez Gene ID
35332
Gene Name
Hormone receptor-like in 38
Gene Aliases
38E.3, 38E.7, CG1864, DHR38, Dhr38, Dmel\CG1864, Dmel_CG1864, HR38, NR4A4, dHR38, dhr38, hr38, l(2)02306
Chromosome Location
Chr.2L: 20683462 - 20714588 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014859
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169553.2 NP_001163024.1 3 - 4 1203 65
NM_057771.4 NP_477119.1 2 - 3 1863 65

View Full Product Details