The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144291.2 | NP_001137763.2 | 1 - 2 | 1172 | 101 |
NM_001169377.1 | NP_001162848.1 | 1 - 2 | 1172 | 101 | |
NM_057891.3 | NP_477239.2 | 1 - 2 | 1172 | 101 |