Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
drongo
Entrez Gene ID
33263
Gene Name
CG3365-PC
Gene Aliases
CG3365, DRONGO, Dmel\CG3365, Dmel_CG3365
Chromosome Location
Chr.2L: 833584 - 851071 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020304
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144291.2 NP_001137763.2 3 - 4 2198 74
NM_001169377.1 NP_001162848.1 4 - 5 2666 74
NM_001169378.2 NP_001162849.1 4 - 5 1823 74
NM_057891.3 NP_477239.2 3 - 4 2198 74
NM_164392.3 NP_722660.1 3 - 4 1355 74
GenBank mRNA AY119526.1 - 3 - 4 1083 74

View Full Product Details