Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mct1
Entrez Gene ID
31198
Gene Name
Monocarboxylate transporter 1
Gene Aliases
CG3456, Dmel\CG3456, Dmel_CG3456, EG:103B4.3, MCT1
Chromosome Location
Chr.X: 2189188 - 2205373 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023549
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297881.1 NP_001284810.1 3 - 4 677 61
NM_057986.4 NP_477334.2 3 - 4 894 61
GenBank mRNA BT010002.1 - 3 - 4 892 61

View Full Product Details