Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- aurB
- Entrez Gene ID
-
34504
- Gene Name
- aurora B
- Gene Aliases
-
ABK, Aur, Aur B, AurB, Aurora B, Aurora-B, CG6620, DmAIRK2, DmAurB, DmAuroraB, Dmel\CG6620, Dmel_CG6620, IAL, IAL1, Ial, ial, ial/AurB
- Chromosome Location
-
Chr.2L: 10856797 - 10858094 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024227
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057988.4 |
NP_477336.1 |
1 - 2 |
248 |
91 |
GenBank mRNA |
AY118562.1 |
- |
1 - 2 |
205 |
91 |
View Full Product Details