Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ph-d
- Entrez Gene ID
-
44889
- Gene Name
- polyhomeotic distal
- Gene Aliases
-
CG3895, DROZFP, Dmel\CG3895, Dmel_CG3895, EG:BACN25G24.3, PH, PH-d, PHD, Ph, Ph-d, PhD, ph, ph-D, ph[D], ph[d], phd, phm
- Chromosome Location
-
Chr.X: 2115727 - 2123286 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004860
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080055.3 |
NP_524794.2 |
1 - 2 |
102 |
81 |
View Full Product Details