Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Spn43Aa
- Entrez Gene ID
-
45041
- Gene Name
- Serpin 43Aa
- Gene Aliases
-
CG12172, Dmel\CG12172, Dmel_CG12172, SER1, Ser1, Spn43 Aa, Spn43A, anon-WO0118057.1
- Chromosome Location
-
Chr.2R: 7147692 - 7149289 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024294
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080066.3 |
NP_524805.1 |
3 - 4 |
1088 |
68 |
GenBank mRNA |
AJ245442.1 |
- |
3 - 4 |
1080 |
68 |
View Full Product Details