Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CIAPIN1
- Entrez Gene ID
-
49424
- Gene Name
- Cytokine induced apoptosis inhibitor 1
- Gene Aliases
-
B, BG:DS00929.9, CG4180, D-CIAPIN1, D1, Dmel\CG4180, Dmel_CG4180, anon-35Ba, br26, l(2)35Bg, l(2)br26
- Chromosome Location
-
Chr.2L: 15036293 - 15038861 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001977
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080199.3 |
NP_524938.1 |
2 - 3 |
720 |
84 |
GenBank mRNA |
AY060402.1 |
- |
2 - 3 |
682 |
84 |
View Full Product Details