Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rdgC
Entrez Gene ID
40224
Gene Name
retinal degeneration C
Gene Aliases
CG13814, CG44746, CG6571, Dmel\CG44746, Dmel_CG44746, DmrdgC-77B, RDGC, RDGC/PP5, RdgC, RdgC 77B
Chromosome Location
Chr.3L: 20248080 - 20265554 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265959
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001370029.1 NP_001356983.1 9 - 10 1429 79
NM_080490.4 NP_536738.3 10 - 11 2278 79
NM_176366.2 NP_788544.2 10 - 11 1916 79
NM_176367.3 NP_788545.3 10 - 11 2091 79
NM_176368.2 NP_788546.1 9 - 10 1606 79

View Full Product Details