Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Stip1
- Entrez Gene ID
-
33202
- Gene Name
- Stress induced phosphoprotein 1
- Gene Aliases
-
CG2720, Dmel\CG2720, Dmel_CG2720, EP(2)0418, HOP, Hop, Sti1, dHop, dSTI1, dSTIP1, dhop, dmSTI1, hop, l(2)k00616
- Chromosome Location
-
Chr.2L: 295122 - 297449 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024352
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058006.4 |
NP_477354.1 |
2 - 3 |
445 |
66 |
GenBank mRNA |
BT021419.1 |
- |
2 - 3 |
400 |
66 |
View Full Product Details