Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BubR1
- Entrez Gene ID
-
35522
- Gene Name
- Bub1-related kinase
- Gene Aliases
-
31/13, BUB1, BUBR1, BcDNA:LD23835, Bub1, CG 7838, CG7838, DmBUB1, DmBubRI, Dmbub1, Dmel\CG7838, Dmel_CG7838, bub1, bubR1, bubr1, dBUB1, l(2)k03113, l(2)k06109, mcl(2)Z1525
- Chromosome Location
-
Chr.2R: 5969099 - 5974385 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0263855
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058034.4 |
NP_477382.1 |
1 - 2 |
432 |
92 |
View Full Product Details