Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JTBR
Entrez Gene ID
38235
Gene Name
CG1935-PA
Gene Aliases
BcDNA:RH07980, CG1935, Dmel\CG1935, Dmel_CG1935, anon-EST:fe2G4, dJTB-like, dJTBR
Chromosome Location
Chr.3L: 1802008 - 1802856 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025820
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274360.1 NP_001261289.1 2 - 2 614 156
NM_001274361.1 NP_001261290.1 2 - 2 624 156
NM_058055.5 NP_477403.1 1 - 1 674 156
GenBank mRNA BT032659.1 - 1 - 1 654 156

View Full Product Details