Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Inos
Entrez Gene ID
35671
Gene Name
myo-inositol-1-phosphate synthase
Gene Aliases
CG11143, Dmel\CG11143, Dmel_CG11143, INOS, bs36h12.y1
Chromosome Location
Chr.2R: 7454922 - 7458874 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025885
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058057.6 NP_477405.1 3 - 4 1208 56
GenBank mRNA AF071103.1 - 3 - 4 1211 56
BT001772.1 - 3 - 4 1198 56

View Full Product Details