Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RhoGAP19D
- Entrez Gene ID
-
33048
- Gene Name
- Rho GTPase activating protein at 19D
- Gene Aliases
-
1412, CG1412, Dmel\CG1412, Dmel_CG1412
- Chromosome Location
-
Chr.X: 20469955 - 20510694 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031118
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134552.6 |
NP_608396.2 |
7 - 8 |
2967 |
57 |
View Full Product Details