Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- pip
- Entrez Gene ID
-
40104
- Gene Name
- pipe
- Gene Aliases
-
CG9614, Dmel\CG9614, Dmel_CG9614, Pipe, l(3)76BDp, pipe-ST2
- Chromosome Location
-
Chr.3L: 19298623 - 19337609 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003089
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176351.4 |
NP_788529.1 |
3 - 4 |
495 |
79 |
View Full Product Details