Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- aurA
- Entrez Gene ID
-
41446
- Gene Name
- aurora A
- Gene Aliases
-
87A7-9/2, AAK, AUR, AURKA, Aur, Aur-A, AurA, CG3068, Dmel\CG3068, Dmel_CG3068, aur, aur-A, aurka, ck10, ck[10], l(3)87Ac, l(3)ck10
- Chromosome Location
-
Chr.3R: 11963124 - 11964826 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000147
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057401.4 |
NP_476749.1 |
2 - 3 |
583 |
102 |
GenBank mRNA |
AY061229.1 |
- |
2 - 3 |
505 |
102 |
View Full Product Details