Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mbc
Entrez Gene ID
42817
Gene Name
myoblast city
Gene Aliases
CG10379, DOCK180, Dm MBC, Dmel\CG10379, Dmel_CG10379, Dock180, MBC, MBC/DOCK180, Mbc, Su(rac)1, dock180
Chromosome Location
Chr.3R: 23781765 - 23801634 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015513
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260338.2 NP_001247267.1 1 - 2 622 79
NM_057796.4 NP_477144.1 1 - 2 622 79
GenBank mRNA AF007805.1 - 1 - 2 607 79

View Full Product Details