Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kkv
Entrez Gene ID
45884
Gene Name
krotzkopf verkehrt
Gene Aliases
Blimp, CG2666, CS-1, CS-1/kkv, ChSB, DmCS-1, DmeCHS1, DmeChSB, Dmel\CG2666, Dmel_CG2666, Kkv, blimp, chitin synthase, l(3)82Fh
Chromosome Location
Chr.3R: 5378093 - 5392866 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001311
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079509.3 NP_524233.1 2 - 3 272 59
NM_169052.3 NP_730928.2 2 - 3 272 59
NM_206430.3 NP_996152.1 2 - 3 272 59

View Full Product Details