Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Irp-1B
- Entrez Gene ID
-
41269
- Gene Name
- Iron regulatory protein 1B
- Gene Aliases
-
144037_at, C-Acon, CG6342, Dmel\CG6342, Dmel_CG6342, IRF, IRP, IRP-1, IRP-1B, IRP1, IRP1B, cACON, cAcon, dIRP, irp-1B, irp1B
- Chromosome Location
-
Chr.3R: 10408618 - 10412869 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024957
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079579.3 |
NP_524303.2 |
9 - 10 |
2644 |
68 |
View Full Product Details