Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TkR86C
Entrez Gene ID
41286
Gene Name
Tachykinin-like receptor at 86C
Gene Aliases
CG6515, DmNTLR, Dmel\CG6515, Dmel_CG6515, NKD, NKD12, NKr, Nkd, TAKR86C, TakR86C, Takr86C
Chromosome Location
Chr.3R: 10737592 - 10751228 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004841
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104271.1 NP_001097741.1 5 - 6 1707 68
NM_079580.3 NP_524304.2 5 - 6 1296 68
GenBank mRNA BT029262.1 - 5 - 6 1707 68

View Full Product Details