Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snk
Entrez Gene ID
41607
Gene Name
snake
Gene Aliases
CG7996, Dmel\CG7996, Dmel_CG7996, SNK, SP26, Snk, c-SP26, cSP26, l(3)87Dg, me(3)4
Chromosome Location
Chr.3R: 13040165 - 13042770 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003450
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104296.2 NP_001097766.1 4 - 5 813 58
NM_079614.3 NP_524338.2 3 - 4 782 58
GenBank mRNA BT024420.1 - 4 - 5 813 58

View Full Product Details