Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mRpS11
Entrez Gene ID
42061
Gene Name
mitochondrial ribosomal protein S11
Gene Aliases
BcDNA:RH70248, CG5184, Dmel\CG5184, Dmel_CG5184, GH1, MRP-S11
Chromosome Location
Chr.3R: 17072732 - 17073795 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038474
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079658.4 NP_524382.1 2 - 3 398 81
GenBank mRNA AY095194.1 - 2 - 3 398 81
DQ062760.1 - 2 - 3 332 81

View Full Product Details