Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or94a
- Entrez Gene ID
-
42711
- Gene Name
- Odorant receptor 94a
- Gene Aliases
-
94D.1, 94a, CG17241, DOR108, Dmel\CG17241, Dmel_CG17241
- Chromosome Location
-
Chr.3R: 22987281 - 22988503 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039033
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079731.1 |
NP_524455.1 |
1 - 2 |
741 |
109 |
View Full Product Details