Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DNApol-epsilon255
- Entrez Gene ID
-
42758
- Gene Name
- DNA polymerase epsilon 255kD subunit
- Gene Aliases
-
6768, BAB17608, CG6768, DNA pol epsilon, DNApol-epsilon, Dmel\CG6768, Dmel_CG6768, Polepsilon, Polepsilon255, Polepsilon255KDa, dpolepsilon, dpolepsilon255, dpolepsilonp255, l(3)pl10, l(3)pl10R, pl[10], pol-epsilon
- Chromosome Location
-
Chr.3R: 23348128 - 23355163 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0264326
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079738.3 |
NP_524462.2 |
1 - 2 |
137 |
64 |
View Full Product Details