Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sym
Entrez Gene ID
40709
Gene Name
Symplekin
Gene Aliases
CG2097, Dmel\CG2097, Dmel_CG2097, Symp, dSymp, symplekin
Chromosome Location
Chr.3R: 5738178 - 5742476 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037371
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141323.2 NP_649580.1 2 - 3 629 79
GenBank mRNA AY118592.1 - 2 - 3 360 79

View Full Product Details