Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osi10b
Entrez Gene ID
40764
Gene Name
Osiris 10b
Gene Aliases
10b_CG15593, CG15593, CG46437, Dmel\CG46437, Dmel_CG46437, Osi10
Chromosome Location
Chr.3R: 6266190 - 6267457 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0286977
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382154.1 NP_001368993.1 2 - 3 621 104

View Full Product Details