Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sowi
Entrez Gene ID
40809
Gene Name
solwind
Gene Aliases
CG15178, Dmel\CG15178, Dmel_CG15178
Chromosome Location
Chr.3R: 6626930 - 6627900 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037460
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141414.2 NP_649671.2 2 - 3 614 82
GenBank mRNA BT022649.1 - 2 - 3 613 82
BT022678.1 - 2 - 3 616 82

View Full Product Details