Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Obp85a
Entrez Gene ID
41006
Gene Name
Odorant-binding protein 85a
Gene Aliases
CG11732, DmelObp85a, Dmel\CG11732, Dmel_CG11732
Chromosome Location
Chr.3R: 8419822 - 8420597 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037589
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347742.1 NP_001334691.1 2 - 3 165 76

View Full Product Details