Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCT7
- Entrez Gene ID
-
41054
- Gene Name
- Chaperonin containing TCP1 subunit 7
- Gene Aliases
-
CG8351, Cct7, Dmel\CG8351, Dmel_CG8351, Q9VHL2_DROME, T-cp1eta, TCP1-eta, TCP1eta, TCPeta, Tcp-1-eta1, Tcp-1eta, tcp-1eta, tcp1eta
- Chromosome Location
-
Chr.3R: 8801564 - 8806234 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037632
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141578.2 |
NP_649835.1 |
4 - 5 |
1524 |
57 |
GenBank mRNA |
AY122201.1 |
- |
4 - 5 |
1496 |
57 |
View Full Product Details