Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cap-H2
Entrez Gene ID
41289
Gene Name
Chromosome associated protein H2
Gene Aliases
CAP-H2, CG14685, CapH2, Dmel\CG14685, Dmel_CG14685, cap-H2, dCap-H2
Chromosome Location
Chr.3R: 10761310 - 10766756 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037831
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141757.2 NP_650014.2 2 - 3 495 78
NM_169358.2 NP_731502.2 2 - 3 495 78
NM_169359.2 NP_731503.2 2 - 3 495 78
NM_206471.2 NP_996193.1 1 - 2 260 78
GenBank mRNA BT001334.1 - 1 - 2 260 78

View Full Product Details