Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dic1
Entrez Gene ID
41640
Gene Name
Dicarboxylate carrier 1
Gene Aliases
BcDNA:GH02431, CG8790, DmDic1p, Dmel\CG8790, Dmel_CG8790
Chromosome Location
Chr.3R: 13364938 - 13369278 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027610
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300384.1 NP_001287313.1 2 - 3 557 73
NM_142022.4 NP_650279.1 2 - 3 557 73
NM_169510.3 NP_731793.1 2 - 3 712 73
GenBank mRNA AF145602.1 - 2 - 3 684 73

View Full Product Details