Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PR-Set7
Entrez Gene ID
41743
Gene Name
PR/SET domain containing protein 7
Gene Aliases
CG3307, Dmel\CG3307, Dmel_CG3307, PR-SET7, PR/SET07, PRSET7, PRSet7, Pr-set-7, PrSet7, SET8, Set7, Set8, dPR-Set7, dSET8, dSet8, l(3)neo41, pr-Set7, pr-set7
Chromosome Location
Chr.3R: 14316038 - 14320494 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011474
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260171.2 NP_001247100.1 2 - 3 419 79
NM_142097.4 NP_650354.1 2 - 3 555 79
NM_169577.3 NP_731900.1 3 - 4 625 79
GenBank mRNA AY102673.1 - 2 - 3 543 79
AY283060.1 - 1 - 2 254 79

View Full Product Details