Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trs33
Entrez Gene ID
41866
Gene Name
TRAPP subunit 33
Gene Aliases
BcDNA:RH37427, CG6196, Dmel\CG6196, Dmel_CG6196, TRAPPC6, Trs33p, dTrs33
Chromosome Location
Chr.3R: 15350345 - 15350995 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266722
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142193.4 NP_650450.1 1 - 2 150 86
GenBank mRNA AY071721.1 - 1 - 2 175 86
KX532055.1 - 1 - 2 75 86

View Full Product Details