Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B9d1
Entrez Gene ID
41888
Gene Name
B9d1-PA
Gene Aliases
B9D1, CG14870, Dmel\CG14870, Dmel_CG14870
Chromosome Location
Chr.3R: 15481906 - 15482904 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038342
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142213.2 NP_650470.1 1 - 2 488 66
GenBank mRNA BT032728.1 - 1 - 2 488 66

View Full Product Details