Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Fbxl7
- Entrez Gene ID
-
41935
- Gene Name
- F-box and leucine-rich repeat protein 7
- Gene Aliases
-
CG4221, Dmel\CG4221, Dmel_CG4221, FBXL7, FbxL7, fbxl7
- Chromosome Location
-
Chr.3R: 15920898 - 15932887 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038385
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142255.3 |
NP_650512.1 |
2 - 3 |
1284 |
52 |
GenBank mRNA |
AY058675.1 |
- |
2 - 3 |
1269 |
52 |
View Full Product Details