Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- psidin
- Entrez Gene ID
-
42389
- Gene Name
- phagocyte signaling impaired
- Gene Aliases
-
CG4845, Dmel\CG4845, Dmel_CG4845, Naa25, ird16, psid
- Chromosome Location
-
Chr.3R: 20021704 - 20026482 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0243511
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142601.2 |
NP_650858.1 |
3 - 4 |
1129 |
78 |
View Full Product Details