Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Obp93a
- Entrez Gene ID
-
42506
- Gene Name
- Odorant-binding protein 93a
- Gene Aliases
-
CG17284, DmelObp93a, Dmel\CG17284, Dmel_CG17284
- Chromosome Location
-
Chr.3R: 21128950 - 21129717 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038859
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142688.1 |
NP_650945.1 |
2 - 3 |
235 |
76 |
View Full Product Details