Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wda
Entrez Gene ID
42750
Gene Name
will decrease acetylation
Gene Aliases
CG4448, Dmel\CG4448, Dmel_CG4448, WDA, WFA, Wda, dWda
Chromosome Location
Chr.3R: 23255070 - 23258087 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039067
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142879.3 NP_651136.1 3 - 4 1840 98
GenBank mRNA AY118904.1 - 3 - 4 1809 98

View Full Product Details