Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SdhD
Entrez Gene ID
42808
Gene Name
Succinate dehydrogenase, subunit D
Gene Aliases
BEST:GH27735, CG10219, DHSD, Dmel\CG10219, Dmel_CG10219
Chromosome Location
Chr.3R: 23759483 - 23760600 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039112
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142924.3 NP_651181.1 1 - 2 154 73
GenBank mRNA AY071388.1 - 1 - 2 151 73
AY071617.1 - 1 - 2 151 73

View Full Product Details