Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- polybromo
- Entrez Gene ID
-
42954
- Gene Name
- CG11375-PA
- Gene Aliases
-
BAF180, Bap180, CG11375, Dmel\CG11375, Dmel_CG11375, PB, POLYBROMO, Pb, Polybromo, anon-WO0140519.158, baf180, bap180
- Chromosome Location
-
Chr.3R: 24799967 - 24805259 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039227
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143031.2 |
NP_651288.1 |
1 - 2 |
1038 |
65 |
View Full Product Details